1H,1H,2H-Perfluoro-1-butene 97%

Supplier: Apollo Scientific
Warning

PC5564-5G PC5564-25G
APOSPC5564-5GEA 292 EUR
APOSPC5564-5G APOSPC5564-25G
1H,1H,2H-Perfluoro-1-butene 97%
1H,1H,2H-Perfluoro-1-butene
Formula: C₄H₃F₅
MW: 146,06 g/mol
Boiling Pt: 44350 °C
Storage Temperature: Ambient
CAS Number: 374-27-6
UN: 3161
ADR: 2.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR