Pentafluoronitrobenzene 98%
Supplier: Apollo Scientific
|
Warning
|
PC5610-25G
PC5610-5G
PC5610-1G
APOSPC5610-25GEA
361
EUR
APOSPC5610-25G
APOSPC5610-5G
APOSPC5610-1G
Pentafluoronitrobenzene 98%
Pentafluoronitrobenzene
Formula:
C₆F₅NO₂ Boiling Pt: 158…161 °C Density: 1,656 g/cm³ (25 °C) Flash Pt: 91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007049 CAS Number: 880-78-4 EINECS: 212-915-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...