Tetraethylammonium fluoride hydrate 98%
Supplier: Apollo Scientific
Synonyms:
Tetrylammonium fluoride hydrate, Tetraethylammonium fluoride xhydrate
PC56243-100G
PC56243-25G
PC56243-5G
APOSPC56243-100GEA
379
EUR
APOSPC56243-100G
APOSPC56243-25G
APOSPC56243-5G
Tetraethylammonium fluoride hydrate 98%
Tetraethylammonium fluoride hydrate
Formula:
C₈H₂₀FN·xH₂O MW: 149.25 g/mol (anhy) Storage Temperature: Ambient |
MDL Number:
MFCD00149990 CAS Number: 98330-04-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...