4-Fluorophenylglyoxal hydrate
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(4-Fluorophenyl)-2-oxoacetaldehyde hydrate, 4-Fluorophenyl(oxo)glyoxal hydrate
PC5633-1G
PC5633-5G
PC5633-10G
APOSPC5633-1GEA
21.9
EUR
APOSPC5633-1G
APOSPC5633-5G
APOSPC5633-10G
4-Fluorophenylglyoxal hydrate
4-Fluorophenylglyoxal hydrate
Formula:
C₈H₇FO₃ MW: 170,140 g/mol Boiling Pt: 115 °C/35mmHg Melting Pt: 80…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD01320770 CAS Number: 447-43-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...