(Pentafluorophenyl)acetonitrile 98%

Supplier: Apollo Scientific
Warning

Synonyms: 2,3,4,5,6-Pentafluorobenzyl cyanide, 2-(Perfluorophenyl)acetonitrile, Pentafluorobenzyl cyanide, (2,3,4,5,6-Pentafluorophenyl)acetonitrile

PC5660-25G PC5660-5G
APOSPC5660-25GEA 288 EUR
APOSPC5660-25G APOSPC5660-5G
(Pentafluorophenyl)acetonitrile 98%
(Pentafluorophenyl)acetonitrile
Formula: (C₆F₅)CH₂CN
MW: 207.1 g/mol
Boiling Pt: 107…111 °C/17mmHg
Density: 1.61 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00013814
CAS Number: 653-30-5
EINECS: 211-498-6
UN: 3276
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR