N-Pentafluorophenyldichloromaleimide 97%
Supplier: Apollo Scientific
|
Warning
|
PC5663E-1G
PC5663E-5G
APOSPC5663E-1GEA
94.4
EUR
APOSPC5663E-1G
APOSPC5663E-5G
N-Pentafluorophenyldichloromaleimide 97%
N-Pentafluorophenyldichloromaleimide
Formula:
C₁₀Cl₂F₅NO₂ Melting Pt: 148…151 °C Storage Temperature: Ambient |
MDL Number:
MFCD00054728 CAS Number: 186958-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...