3-Chloro-4-((3-fluorobenzyl)oxy)aniline 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Chloro-4-(3-fluorobenzyloxy)aniline
PC5666-25G
PC5666-5G
APOSPC5666-25GEA
87.3
EUR
APOSPC5666-25G
APOSPC5666-5G
3-Chloro-4-((3-fluorobenzyl)oxy)aniline 98%
3-Chloro-4-((3-fluorobenzyl)oxy)aniline
Formula:
C₁₃H₁₁ClFNO MW: 251,69 g/mol Melting Pt: 82…84 °C Storage Temperature: Ambient |
MDL Number:
MFCD06809822 CAS Number: 202197-26-0 UN: 3077 ADR: 9+EHS,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...