5-Fluorosalicylaldehyde
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
5-Fluoro-2-hydroxybenzaldehyde
PC5683-100G
PC5683-25G
PC5683-5G
APOSPC5683-100GEA
176
EUR
APOSPC5683-100G
APOSPC5683-25G
APOSPC5683-5G
5-Fluorosalicylaldehyde
5-Fluorosalicylaldehyde
Formula:
C₇H₅FO₂ MW: 140,11 g/mol Boiling Pt: 56 °C/0,75mmHg Melting Pt: 82…85 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD01090997 CAS Number: 347-54-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...