Sodium-3-fluorobenzoate 95%
Supplier: Apollo Scientific
|
Warning
|
PC5732-100G
PC5732-25G
PC5732-500G
APOSPC5732-100GEA
58
EUR
APOSPC5732-100G
APOSPC5732-25G
APOSPC5732-500G
Sodium-3-fluorobenzoate 95%
Sodium-3-fluorobenzoate
Formula:
C₇H₄FO₂Na Storage Temperature: Ambient |
MDL Number:
MFCD03788550 CAS Number: 499-57-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...