2,2,3,3,3-Pentafluoropropylamine 97%
Supplier: Apollo Scientific
|
Danger
|
PC5786-5G
PC5786-1G
APOSPC5786-5GEA
126
EUR
APOSPC5786-5G
APOSPC5786-1G
2,2,3,3,3-Pentafluoropropylamine 97%
2,2,3,3,3-Pentafluoropropylamine
Formula:
C₃H₄F₅N Boiling Pt: 49…50 °C Density: 1,4 g/cm³ (25 °C) Flash Pt: >100 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042459 CAS Number: 422-03-7 UN: 2735 ADR: 8,III |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...