2,2,3,3,3-Pentafluoropropyl difluoromethyl ether 97%
Supplier: Apollo Scientific
Synonyms:
Difluoromethyl 2,2,3,3,3-pentafluoropropyl ether
PC5786F-1G
PC5786F-5G
APOSPC5786F-1GEA
261
EUR
APOSPC5786F-1G
APOSPC5786F-5G
2,2,3,3,3-Pentafluoropropyl difluoromethyl ether 97%
2,2,3,3,3-Pentafluoropropyl difluoromethyl ether
Formula:
C₄H₃F₇O Boiling Pt: 46 °C Density: 1,46 g/cm³ (25 °C) Storage Temperature: Ambient |
CAS Number:
56860-81-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...