2-Fluoro-5-(trifluoromethoxy)aniline 99%
Supplier: Apollo Scientific
|
Danger
|
PC5900-1G
PC5900-25G
PC5900-5G
APOSPC5900-1GEA
30.6
EUR
APOSPC5900-1G
APOSPC5900-25G
APOSPC5900-5G
2-Fluoro-5-(trifluoromethoxy)aniline 99%
2-Fluoro-5-(trifluoromethoxy)aniline
Formula:
C₇H₅F₄NO MW: 195,12 g/mol Boiling Pt: 64…65 °C/15mmHg Storage Temperature: Ambient |
MDL Number:
MFCD09832254 CAS Number: 116369-23-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...