2-Fluoro-p-toluic acid
Supplier: Apollo Scientific
|
Warning
|
PC5934-5G
PC5934-100G
PC5934-25G
APOSPC5934-5GEA
21.9
EUR
APOSPC5934-5G
APOSPC5934-100G
APOSPC5934-25G
2-Fluoro-p-toluic acid
2-Fluoro-p-toluic acid
Formula:
C₈H₇FO₂ MW: 154,14 g/mol Melting Pt: 185…189 °C Storage Temperature: Ambient |
MDL Number:
MFCD03092906 CAS Number: 7697-23-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...