1-Bromoperfluoroheptane 97%

Supplier: Apollo Scientific
Warning

PC6050-25G PC6050-5G
APOSPC6050-25GEA 203 EUR
APOSPC6050-25G APOSPC6050-5G
1-Bromoperfluoroheptane 97%
1-Bromoperfluoroheptane
Formula: BrF₂C(CF₂)₅CF₃
Boiling Pt: 117…119 °C
Density: 1,894 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00013570
CAS Number: 375-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR