1-Iodoperfluoroheptane 97%

Supplier: Apollo Scientific
Warning

PC6060-1G PC6060-25G PC6060-5G PC6060-100G
APOSPC6060-1GEA 65.7 EUR
APOSPC6060-1G APOSPC6060-25G APOSPC6060-5G APOSPC6060-100G
1-Iodoperfluoroheptane 97%
1-Iodoperfluoroheptane
Formula: C₇F₁₅I
Boiling Pt: 137…138 °C
Density: 2,01 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00013712
CAS Number: 335-58-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR