Perfluorohexadecane
Supplier: Apollo Scientific
|
Warning
|
PC6064-1G
PC6064-5G
APOSPC6064-1GEA
93.4
EUR
APOSPC6064-1G
APOSPC6064-5G
Perfluorohexadecane
Perfluorohexadecane
Formula:
C₁₆F₃₄ Boiling Pt: 239…240 °C Melting Pt: 125…126 °C Storage Temperature: Ambient |
CAS Number:
355-49-7 EINECS: 206-588-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...