5-Bromo-3-fluoro-2-pyridine carboxylic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 5-Bromo-3-fluoropicolinic acid, 5-Bromo-3-fluoropyridine-2-carboxylic acid

PC6074-1G PC6074-5G
APOSPC6074-1GEA 21.9 EUR
APOSPC6074-1G APOSPC6074-5G
5-Bromo-3-fluoro-2-pyridine carboxylic acid 98%
5-Bromo-3-fluoro-2-pyridine carboxylic acid
Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Melting Pt: 175…180 °C
Storage Temperature: Ambient
MDL Number: MFCD07437865
CAS Number: 669066-91-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR