Perfluoro(4-methylmorpholine) 98%

Supplier: Apollo Scientific
Warning

PC6122-10G PC6122-25G PC6122-5G PC6122-100G
APOSPC6122-10GEA 0 EUR
APOSPC6122-10G APOSPC6122-25G APOSPC6122-5G APOSPC6122-100G
Perfluoro(4-methylmorpholine) 98%
Perfluoro(4-methylmorpholine)
Formula: C₅F₁₁NO
MW: 299,04 g/mol
Boiling Pt: 50 °C
Density: 1,7 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00042442
CAS Number: 382-28-5
EINECS: 206-841-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR