1H,1H-Pentatriacontafluoro-1-octadecanol 95%

Supplier: Apollo Scientific
Warning

PC6137-1G PC6137-250MG
APOSPC6137-1GEA 348 EUR
APOSPC6137-1G APOSPC6137-250MG
1H,1H-Pentatriacontafluoro-1-octadecanol 95%
1H,1H-Pentatriacontafluoro-1-octadecanol
Formula: C₁₈H₃F₃₅O
Boiling Pt: 211 °C/100mmHg
Melting Pt: 152…156 °C
Density: 0,936 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236627
CAS Number: 242142-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR