Nonafluorovaleric acid
Supplier: Apollo Scientific
|
Danger
|
PC6184-25G
PC6184-5G
APOSPC6184-25GEA
108
EUR
APOSPC6184-25G
APOSPC6184-5G
Nonafluorovaleric acid
Nonafluorovaleric acid
Formula:
C₅HF₉O₂ Boiling Pt: 82 °C/85mmHg Density: 1,713 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00040211 CAS Number: 2706-90-3 EINECS: 220-300-7 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...