α,α,α,2,5-Pentafluorotoluene 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,5-Difluorobenzotrifluoride
PC6199-1G
PC6199-5G
APOSPC6199-1GEA
21.9
EUR
APOSPC6199-1G
APOSPC6199-5G
α,α,α,2,5-Pentafluorotoluene 97%
α,α,α,2,5-Pentafluorotoluene
Formula:
C₇H₃F₅ MW: 182,09 g/mol Boiling Pt: 107…108 °C Melting Pt: –14 °C Density: 1,424 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042494 CAS Number: 393-38-4 |
Learn more
![](/m-ie.vwr.com/en_IE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...