α,α,α,3,5-Pentafluorotoluene 99+%

Supplier: Apollo Scientific
Warning

Synonyms: 3,5-Difluorobenzotrifluoride

PC6205-1G PC6205-5G
APOSPC6205-1GEA 108 EUR
APOSPC6205-1G APOSPC6205-5G
α,α,α,3,5-Pentafluorotoluene 99+%
α,α,α,3,5-Pentafluorotoluene
Formula: C₇H₃F₅
Boiling Pt: 94…95 °C
Melting Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD09263729
CAS Number: 401-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR