Perfluorotridecane 98%

Supplier: Apollo Scientific
Warning

PC6220A-25G PC6220A-5G
APOSPC6220A-25GEA 439 EUR
APOSPC6220A-25G APOSPC6220A-5G
Perfluorotridecane 98%
Perfluorotridecane
Formula: C₁₃F₂₈
MW: 688.1 g/mol
Boiling Pt: 193…196 °C
Melting Pt: 88…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00236715
CAS Number: 376-03-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR