Silver hydrogen fluoride 99%
Supplier: Apollo Scientific
|
Danger
|
PC6350-5G
APOSPC6350-5GEA
0
EUR
APOSPC6350-5G
Silver hydrogen fluoride 99%
Silver hydrogen fluoride
Formula:
AgF₂H MW: 146,87 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00042145 CAS Number: 12249-52-4 UN: 1740 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...