2-Fluoro-6-methoxybenzylamine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
(2-Fluoro-6-methoxyphenyl)methanamine
PC6359-1G
PC6359-250MG
APOSPC6359-1GEA
88.7
EUR
APOSPC6359-1G
APOSPC6359-250MG
2-Fluoro-6-methoxybenzylamine
2-Fluoro-6-methoxybenzylamine
Formula:
C₈H₁₀FNO MW: 155,1695 g/mol Boiling Pt: 76…77 °C/3,2mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00042292 CAS Number: 150517-75-2 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...