4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde

Supplier: Apollo Scientific
Warning

PC6379-1G PC6379-5G
APOSPC6379-1GEA 129 EUR
APOSPC6379-1G APOSPC6379-5G
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde
4-{[3-(Trifluoromethyl)pyridin-2-yl]oxy}benzaldehyde
Formula: C₁₃H₈F₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD10703516
CAS Number: 1086379-05-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR