2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%

Supplier: Apollo Scientific
Danger

PC6431-1G PC6431-5G
APOSPC6431-1GEA 21.9 EUR
APOSPC6431-1G APOSPC6431-5G
2-Hydrazino-4-(trifluoromethyl)pyrimidine 97%
2-Hydrazino-4-(trifluoromethyl)pyrimidine
Formula: C₅H₅F₃N₄
Melting Pt: 99…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00115112
CAS Number: 197305-97-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR