Sodium trifluoroacetate 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Sodium 2,2,2-trifluoroacetate
PC6570-100G
PC6570-25G
PC6570-500G
APOSPC6570-100GEA
49.3
EUR
APOSPC6570-100G
APOSPC6570-25G
APOSPC6570-500G
Sodium trifluoroacetate 98%
Sodium trifluoroacetate
Formula:
C₂F₃NaO₂ MW: 136.01 g/mol Melting Pt: 205…207 °C (dec.) Density: 1.49 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013217 CAS Number: 2923-18-4 EINECS: 220-879-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...