o,α,α,α-Tetrafluoroanisole 98%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
α,α,α,2-Tetrafluoroanisole, 2-(Trifluoromethoxy)fluorobenzene, 1-Fluoro-2-(trifluoromethoxy)benzene
PC6573-100G
PC6573-25G
PC6573-5G
APOSPC6573-100GEA
165
EUR
APOSPC6573-100G
APOSPC6573-25G
APOSPC6573-5G
o,α,α,α-Tetrafluoroanisole 98%
o,α,α,α-Tetrafluoroanisole
Formula:
C₇H₄F₄O MW: 180,1 g/mol Boiling Pt: 105…106 °C Density: 1,3 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00236322 CAS Number: 2106-18-5 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...