2,4-Diamino-6-[3-(trifluoromethyl)phenyl]-1,3,5-triazine

Supplier: Apollo Scientific
Danger

Synonyms: 6-(3-(Trifluoromethyl)phenyl)-1,3,5-triazine-2,4-diamine

PC6632-1G PC6632-25G PC6632-5G
APOSPC6632-1GEA 74.1 EUR
APOSPC6632-1G APOSPC6632-25G APOSPC6632-5G
2,4-Diamino-6-[3-(trifluoromethyl)phenyl]-1,3,5-triazine
2,4-Diamino-6-[3-(trifluoromethyl)phenyl]-1,3,5-triazine
Formula: C₁₀H₈F₃N₅
MW: 255.204 g/mol
Boiling Pt: 333…334 °C
Melting Pt: 207…211 °C
Storage Temperature: Ambient
MDL Number: MFCD00268708
CAS Number: 30508-78-2
UN: 2811
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR