1-Fluorotetradecane 97%
Supplier: Apollo Scientific
|
Warning
|
PC6660-1G
PC6660-5G
APOSPC6660-1GEA
58
EUR
APOSPC6660-1G
APOSPC6660-5G
1-Fluorotetradecane 97%
1-Fluorotetradecane
Formula:
C₁₄H₂₉F Boiling Pt: 119…121 °C/3mmHg Density: 0,828 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00042114 CAS Number: 73180-09-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...