Ethyl iododifluoroacetate 97%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2,2-difluoro-2-iodoacetate, Ethyl difluoro(iodo)acetate

PC6688-25G PC6688-5G PC6688-1G
APOSPC6688-25GEA 698 EUR
APOSPC6688-25G APOSPC6688-5G APOSPC6688-1G
Ethyl iododifluoroacetate 97%
Ethyl iododifluoroacetate
Formula: C₄H₅F₂IO₂
MW: 249,9826 g/mol
Boiling Pt: 65 °C/2mmHg
Density: 1,4 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00798135
CAS Number: 7648-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR