2,3,5,6-Tetrafluoroanisole 97%
Supplier: Apollo Scientific
|
Warning
|
PC6700-25G
PC6700-5G
APOSPC6700-25GEA
147
EUR
APOSPC6700-25G
APOSPC6700-5G
2,3,5,6-Tetrafluoroanisole 97%
2,3,5,6-Tetrafluoroanisole
Formula:
C₇H₄F₄O MW: 180,1 g/mol Boiling Pt: 144…145 °C Density: 1,293 g/cm³ (25 °C) Flash Pt: 41 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000311 CAS Number: 2324-98-3 EINECS: 219-037-0 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...