1,2,3,4-Tetrafluorobenzene 98%
Supplier: Apollo Scientific
|
Danger
|
PC6710-100G
PC6710-25G
PC6710-5G
APOSPC6710-100GEA
313
EUR
APOSPC6710-100G
APOSPC6710-25G
APOSPC6710-5G
1,2,3,4-Tetrafluorobenzene 98%
1,2,3,4-Tetrafluorobenzene
Formula:
C₆H₂F₄ MW: 150,08 g/mol Boiling Pt: 94…96 °C Melting Pt: –42 °C Density: 1,4 g/cm³ (25 °C) Flash Pt: 20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000285 CAS Number: 551-62-2 EINECS: 208-997-6 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...