2,3,5-Trifluoroisonicotinic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2,3,5-Trifluoropyridine-4-carboxylic acid
PC6737-1G
PC6737-5G
APOSPC6737-1GEA
139
EUR
APOSPC6737-1G
APOSPC6737-5G
2,3,5-Trifluoroisonicotinic acid
2,3,5-Trifluoroisonicotinic acid
Formula:
C₆H₂F₃NO₂ MW: 177,08 g/mol Melting Pt: 106…112 °C Storage Temperature: Ambient |
MDL Number:
MFCD03840205 CAS Number: 675602-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...