Tetrafluorophthalonitrile 97%
Supplier: Apollo Scientific
|
Warning
|
PC6860-1G
PC6860-100G
PC6860-10G
PC6860-25G
APOSPC6860-1GEA
14.7
EUR
APOSPC6860-1G
APOSPC6860-100G
APOSPC6860-10G
APOSPC6860-25G
Tetrafluorophthalonitrile 97%
Tetrafluorophthalonitrile
Formula:
C₈F₄N₂ Melting Pt: 81…86 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001774 CAS Number: 1835-65-0 EINECS: 217-400-8 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...