Glycidyl-2,2,3,3-tetrafluoropropyl ether 97%
Supplier: Apollo Scientific
|
Danger
|
PC6867H-25G
PC6867H-5G
PC6867H-1G
APOSPC6867H-25GEA
275
EUR
APOSPC6867H-25G
APOSPC6867H-5G
APOSPC6867H-1G
Glycidyl-2,2,3,3-tetrafluoropropyl ether 97%
Glycidyl-2,2,3,3-tetrafluoropropyl ether
Formula:
C₆H₈O₂F₄ MW: 188,12 g/mol Boiling Pt: 50…52 °C Density: 1,327 g/cm³ (25 °C) Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00054682 CAS Number: 19932-26-4 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...