Tetrafluorosuccinamide 98%

Supplier: Apollo Scientific
Warning

PC6870-1G PC6870-25G PC6870-5G
APOSPC6870-1GEA 61.3 EUR
APOSPC6870-1G APOSPC6870-25G APOSPC6870-5G
Tetrafluorosuccinamide 98%
Tetrafluorosuccinamide
Formula: C₄H₄F₄N₂O₂
MW: 188.08 g/mol
Melting Pt: 259…262 °C
Storage Temperature: Ambient
MDL Number: MFCD00039772
CAS Number: 377-37-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR