3-Fluoro-4-formylphenylboronic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-4-formylbenzeneboronic acid

PC6911-100G PC6911-25G PC6911-5G
APOSPC6911-100GEA 593 EUR
APOSPC6911-100G APOSPC6911-25G APOSPC6911-5G
3-Fluoro-4-formylphenylboronic acid 98%
3-Fluoro-4-formylphenylboronic acid
Formula: C₇H₆BFO₃
MW: 167,93 g/mol
Melting Pt: 240 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02093051
CAS Number: 248270-25-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR