2-Fluoro-6-iodobenzoyl chloride 95%
Supplier: Apollo Scientific
|
Danger
|
PC6949-1G
PC6949-5G
APOSPC6949-1GEA
21.9
EUR
APOSPC6949-1G
APOSPC6949-5G
2-Fluoro-6-iodobenzoyl chloride 95%
2-Fluoro-6-iodobenzoyl chloride
Formula:
C₇H₃ClFIO Boiling Pt: 97…99 °C/5mmHg Density: 1,956 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD03094410 CAS Number: 111771-12-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...