[6-(Trifluoromethyl)-3-pyridinyl]methanol 95+%

Supplier: Apollo Scientific
Danger

PC6985-1G PC6985-5G
APOSPC6985-1GEA 139 EUR
APOSPC6985-1G APOSPC6985-5G
[6-(Trifluoromethyl)-3-pyridinyl]methanol 95+%
[6-(Trifluoromethyl)-3-pyridinyl]methanol
Formula: C₇H₆F₃NO
Boiling Pt: 100…102 °C/0,75mmHg
Density: 1,394 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01862648
CAS Number: 386704-04-7
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR