4,5,7-Trichloro-2-(trifluoromethyl)quinoline
Supplier: Apollo Scientific
|
Warning
|
PC7087E-1G
PC7087E-5G
APOSPC7087E-1GEA
34.8
EUR
APOSPC7087E-1G
APOSPC7087E-5G
4,5,7-Trichloro-2-(trifluoromethyl)quinoline
4,5,7-Trichloro-2-(trifluoromethyl)quinoline
Formula:
C₁₀H₃Cl₃F₃N Melting Pt: 111…114 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153279 CAS Number: 175203-41-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...