2-Chloro-5-fluoropyrimidine 98%

Supplier: Apollo Scientific
Danger

PC7143-100G PC7143-25G PC7143-5G
APOSPC7143-100GEA 190 EUR
APOSPC7143-100G APOSPC7143-25G APOSPC7143-5G
2-Chloro-5-fluoropyrimidine 98%
2-Chloro-5-fluoropyrimidine
Formula: C₄H₂ClFN₂
MW: 132,52 g/mol
Boiling Pt: 149…162 °C
Density: 1,073 g/cm³ (25 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD03788197
CAS Number: 62802-42-0
UN: 3267
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR