Methyl-5-fluoro-2-nitrobenzoate 98%
Supplier: Apollo Scientific
|
Warning
|
PC7145-25G
PC7145-5G
PC7145-1G
APOSPC7145-25GEA
143
EUR
APOSPC7145-25G
APOSPC7145-5G
APOSPC7145-1G
Methyl-5-fluoro-2-nitrobenzoate 98%
Methyl 5-fluoro-2-nitrobenzoate
Formula:
C₈H₆FNO₄ MW: 199,14 g/mol Boiling Pt: 115 °C Melting Pt: 35…37 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03425619 CAS Number: 393-85-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...