2-Bromo-N-(2,4-difluorophenyl)benzamide 98%

Supplier: Apollo Scientific

PC7146-1G PC7146-5G
APOSPC7146-1GEA 87.3 EUR
APOSPC7146-1G APOSPC7146-5G
2-Bromo-N-(2,4-difluorophenyl)benzamide 98%
2-Bromo-N-(2,4-difluorophenyl)benzamide
Formula: C₁₃H₈BrF₂NO
Melting Pt: 110…113 °C
Storage Temperature: Ambient
MDL Number: MFCD00751577
CAS Number: 314025-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR