2-Fluoro-6-formylpyridine 95+%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Fluoropicolinaldehyde, 6-Fluoropyridine-2-carboxaldehyde

PC7169-1G PC7169-10G PC7169-5G
APOSPC7169-1GEA 87.3 EUR
APOSPC7169-1G APOSPC7169-10G APOSPC7169-5G
2-Fluoro-6-formylpyridine 95+%
2-Fluoro-6-formylpyridine
Formula: C₆H₄FNO
MW: 125,1 g/mol
Flash Pt: 70 °C
Storage Temperature: Ambient
MDL Number: MFCD03095149
CAS Number: 208110-81-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR