4-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%
Supplier: Apollo Scientific
Synonyms:
4-Fluoro-2-(trifluoromethyl)benzeneboronic acid, (4-Fluoro-2-(trifluoromethyl)phenyl)boronic acid
PC7175-10G
PC7175-5G
APOSPC7175-10GEA
85.8
EUR
APOSPC7175-10G
APOSPC7175-5G
4-Fluoro-2-(trifluoromethyl)phenylboronic acid 97%
4-Fluoro-2-(trifluoromethyl)phenylboronic acid
Formula:
C₇H₅BF₄O₂ MW: 207,92 g/mol Melting Pt: 183…186 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD08062379 CAS Number: 182344-16-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...