2',3',4'-Trifluoroacetophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(2,3,4-Trifluorophenyl)ethanone, 1-(2,3,4-Trifluorophenyl)ethan-1-one
PC7202D-5G
PC7202D-1G
PC7202D-25G
APOSPC7202D-5GEA
55.3
EUR
APOSPC7202D-5G
APOSPC7202D-1G
APOSPC7202D-25G
2',3',4'-Trifluoroacetophenone 98%
2',3',4'-Trifluoroacetophenone
Formula:
C₈H₅F₃O MW: 174,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061231 CAS Number: 243448-15-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...