2',4',6'-Trifluoroacetophenone 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
1-(2,4,6-Trifluorophenyl)ethanone, 2,4,6-Trifluoroacetophenone, 1-(2,4,6-Trifluorophenyl)ethan-1-one
PC7202K-5G
PC7202K-100G
PC7202K-25G
APOSPC7202K-5GEA
26.1
EUR
APOSPC7202K-5G
APOSPC7202K-100G
APOSPC7202K-25G
2',4',6'-Trifluoroacetophenone 97%
2',4',6'-Trifluoroacetophenone
Formula:
C₈H₅F₃O MW: 174,12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00061194 CAS Number: 51788-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...