3,4,5-Trifluorobenzoic acid 98%

Supplier: Apollo Scientific
Warning

PC7283-25G PC7283-100G PC7283-500G
APOSPC7283-25GEA 29.2 EUR
APOSPC7283-25G APOSPC7283-100G APOSPC7283-500G
3,4,5-Trifluorobenzoic acid 98%
3,4,5-Trifluorobenzoic acid
Formula: C₇H₃F₃O₂
MW: 176,09 g/mol
Melting Pt: 97…99 °C
Storage Temperature: Ambient
MDL Number: MFCD00074962
CAS Number: 121602-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR